Learn More
Teknova 1M HEPES, pH 6.8. 1000mL, Sterile 2 pack.
SDP
Catalog No. 502387495
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
Catalog No. 50-238-7495 Supplier Teknova Supplier No. H10262PK
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.