Learn More
Teknova 1M HEPES, pH 7.4. 1000mL, Sterile.
SDP
Catalog No. NC0470071
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
This item is not returnable. View return policy
Catalog No. NC0470071 Supplier Teknova Supplier No. H1030
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.