Learn More
Bioworld HEPES, 1M, pH 7.4, Sterile, 250 mL
Catalog No. NC0689154
Encompass_Preferred
HEPES BufferAn N-substituted aminosulfonic acid buffer with an optimal buffering pH range of 6.0 to 8.5 Technical Information:Biotechnology GradeDNase(P/F): NoneProtease(P/F): NoneRNase(P/F): NoneAppearance: LiquidBoiling Point : 210-215 °CDensity : 1.33 g/cm3 (Predicted)Melting Point: 234 °C (dec.)PH Value: 7.4RTECS Number: TL6809000HS Code:2933.59.95
Catalog No. NC0689154 Supplier Bioworld Supplier No. 408200081
HEPES BufferAn N-substituted aminosulfonic acid buffer with an optimal buffering pH range of 6.0 to 8.5 Technical Information:Biotechnology GradeDNase(P/F): NoneProtease(P/F): NoneRNase(P/F): NoneAppearance: LiquidBoiling Point : 210-215 °CDensity : 1.33 g/cm3 (Predicted)Melting Point: 234 °C (dec.)PH Value: 7.4RTECS Number: TL6809000HS Code:2933.59.95