Learn More
Teknova 1M HEPES, pH 7.5, 200L Bag, Sterile.
SDP
Catalog No. 502745849
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
Catalog No. 50-274-5849 Supplier Teknova Supplier No. 21361
Add to Cart
Add to Cart
Request a Quote
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.