Learn More
Teknova 1M HEPES, pH 7.5, 20L Bag, Sterile. 4 Pack.
SDP
Catalog No. 502745865
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
Catalog No. 50-274-5865 Supplier Teknova Supplier No. 213524PK
Add to Cart
Add to Cart
Request a Quote
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.