Learn More
Teknova 1M HEPES, pH 8.0. 1000mL, Sterile
SDP
Catalog No. NC0661864
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
This item is not returnable. View return policy
Catalog No. NC0661864 Supplier Teknova Supplier No. H1090
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.