Learn More
Teknova 1M HEPES Solution, pH 6.5. 1000 mL, Sterile.
SDP
Catalog No. 502388017
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
Catalog No. 50-238-8017 Supplier Teknova Supplier No. H1021
Add to Cart
Add to Cart
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.