Learn More
Teknova 1M Imidazole, pH 8.0. 1000mL, Sterile.
SDP
Catalog No. 501907927
Encompass
Imidazole is a small organic compound that is used as a buffer in protein purification to prevent changes in pH, helping to maintain stability of products. It is also used as an antifungal agent and has a wide range of other uses in the food, beverage, and pharmaceutical industries.
Catalog No. 50-190-7927 Supplier Teknova Supplier No. I6980
Imidazole is a small organic compound that is used as a buffer in protein purification to prevent changes in pH, helping to maintain stability of products. It is also used as an antifungal agent and has a wide range of other uses in the food, beverage, and pharmaceutical industries.