Learn More
Boston Bioproducts Inc Manganese Chloride (1 M) - 100ml
Catalog No. NC0391433
Encompass
The 1M Manganese Chloride Solution is an aqueous source of manganese ions, which are involved in numerous biological processes. In biochemistry, manganese chloride is frequently used as a catalyst or co-factor in enzymatic reactions, particularly those involving the synthesis and degradation of carbohydrates and fats. In laboratory settings, manganese ions are often necessary for polymerase chain reactions (PCR), aiding in the replication of DNA and RNA polymerase I and II activity.
Catalog No. NC0391433 Supplier Boston Bioproducts Inc Supplier No. MT220100ML
The 1M Manganese Chloride Solution is an aqueous source of manganese ions, which are involved in numerous biological processes. In biochemistry, manganese chloride is frequently used as a catalyst or co-factor in enzymatic reactions, particularly those involving the synthesis and degradation of carbohydrates and fats. In laboratory settings, manganese ions are often necessary for polymerase chain reactions (PCR), aiding in the replication of DNA and RNA polymerase I and II activity.