Learn More
Teknova 1M MES, pH 5.76. 1000mL, Sterile 2 pack.
SDP
Catalog No. 502387592
Encompass
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.
Catalog No. 50-238-7592 Supplier Teknova Supplier No. M24002PK
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.