Learn More
Molecular Dimensions 1M MIB pH 4.0 and pH 10.0
SDP
Catalog No. NC1410864
Encompass
100 mL bottle of 1M sodium Malonate dibasic monohydrate, Imidazole, Boric acid pH 10.0 and 100 mL bottle of 1M sodium Malonate dibasic monohydrate, Imidazole, Boric acid pH 4.0.
Catalog No. NC1410864 Supplier Molecular Dimensions Supplier No. MD260
100 mL bottle of 1M sodium Malonate dibasic monohydrate, Imidazole, Boric acid pH 10.0 and 100 mL bottle of 1M sodium Malonate dibasic monohydrate, Imidazole, Boric acid pH 4.0.