Learn More
Molecular Dimensions 1M MMT Buffer pH 4 and pH 9
SDP
Catalog No. NC1399458
Encompass
100 mL bottle of 1M DL-Malic acid, MES monohydrate, Tris pH 9.0 and 100 mL bottle of 1M DL-Malic acid, MES monohydrate, Tris pH 4.0
Catalog No. NC1399458 Supplier Molecular Dimensions Supplier No. MD262
100 mL bottle of 1M DL-Malic acid, MES monohydrate, Tris pH 9.0 and 100 mL bottle of 1M DL-Malic acid, MES monohydrate, Tris pH 4.0