Learn More
Teknova 1M Potassium Chloride Solution. 250mL, Sterile. 8 pack.
SDP
Catalog No. 502387922
Encompass
Potassium chloride solution, also known as KCl, is a salt that has a wide range of applications in molecular biology, protein chemistry and biochemical research. It is a key component of many biochemical reactions, including the synthesis of proteins and DNA. In addition, KCl is a popular reagent for the purification of proteins and nucleic acids.
Catalog No. 50-238-7922 Supplier Teknova Supplier No. P03208PK
Potassium chloride solution, also known as KCl, is a salt that has a wide range of applications in molecular biology, protein chemistry and biochemical research. It is a key component of many biochemical reactions, including the synthesis of proteins and DNA. In addition, KCl is a popular reagent for the purification of proteins and nucleic acids.