Learn More
Teknova 1M Sodium Citrate, pH 6.0. 4 L, Sterile 2 Pack.
SDP
Catalog No. 502387769
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-238-7769 Supplier Teknova Supplier No. S84642PK
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.