Learn More
Teknova 1M Sodium Phosphate Buffer, pH 6.5, 1000mL, Sterile.
SDP
Catalog No. 502388020
Encompass
Sodium phosphate is a versatile chemical compound that has a wide range of uses. In molecular biology, it can be used to extract DNA and RNA from cells. In protein chemistry, it can be used to purify proteins. And in biochemical applications, it can be used to study enzyme activity. Sodium phosphate is also an effective buffering agent, making it an ideal solution for use in laboratory and industrial settings. The buffer range is 5.8 to 8.0.
Catalog No. 50-238-8020 Supplier Teknova Supplier No. S0276
Sodium phosphate is a versatile chemical compound that has a wide range of uses. In molecular biology, it can be used to extract DNA and RNA from cells. In protein chemistry, it can be used to purify proteins. And in biochemical applications, it can be used to study enzyme activity. Sodium phosphate is also an effective buffering agent, making it an ideal solution for use in laboratory and industrial settings. The buffer range is 5.8 to 8.0.