Learn More
Teknova 1M Sodium Citrate. pH 6.0, 1000mL. Sterile.
SDP
Catalog No. NC3306517
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. NC3306517 Supplier Teknova Supplier No. S8460
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.