Learn More
Teknova 1M Sorbitol. 1000mL, Sterile.
SDP
Catalog No. NC9838979
Encompass
Sugar is a carbohydrate that serves as a carbon source for microorganisms and can be used to test for the presence of enzymes or other proteins.
Catalog No. NC9838979 Supplier Teknova Supplier No. S1001
Sugar is a carbohydrate that serves as a carbon source for microorganisms and can be used to test for the presence of enzymes or other proteins.