Learn More
Molecular Dimensions 1M SPG pH 4.0 and pH 10.0 | 2 x 100 ml
SDP
Catalog No. NC1875199
Encompass
Buffers not only control the pH of crystallization but also contribute to the chemistry of the reagent with the component buffering ions and chemical species. Each pair from this series of broad-range buffers permits a wide range of pH values to be set up, thus allowing the pH to be changed without altering the buffer chemistry. These special buffers are ideal for applications where the number of stock solutions that can be used is limited, such as with most liquid handling robots. These buffers are also found in PACT premier™. They are provided as 2 x 100 mL untitrated buffer solutions at the pH's specified.
Catalog No. NC1875199 Supplier Molecular Dimensions Supplier No. MD259
Buffers not only control the pH of crystallization but also contribute to the chemistry of the reagent with the component buffering ions and chemical species. Each pair from this series of broad-range buffers permits a wide range of pH values to be set up, thus allowing the pH to be changed without altering the buffer chemistry. These special buffers are ideal for applications where the number of stock solutions that can be used is limited, such as with most liquid handling robots. These buffers are also found in PACT premier™. They are provided as 2 x 100 mL untitrated buffer solutions at the pH's specified.