Learn More
Quality Biological Inc Tris HCl (1 M), pH 7.5, 100 mL
SDP
Catalog No. 507517610
Encompass_Preferred
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. ¿ These solutions of Tris Hydrochloride provide convenient and highly purified solutions. ¿ Applications: Tris solutions are formulated into working concentrations for preparing the stacking and resolving portions of native or SDS-PAGE gels using discontinuous buffer systems according to Laemmli (1970) or Ornstein and Davis (1959). ¿ Use Tris-HCl 1M pH 7.0-7.5 for DEAE Dextran-mediated transfection of non-adherent and adherent cells. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase and Protease Tested ¿ pH Range: 7.5 ¿ 0.1 ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-751-7610 Supplier Quality Biological Inc Supplier No. 351006721EA
Add to Cart
Add to Cart
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. ¿ These solutions of Tris Hydrochloride provide convenient and highly purified solutions. ¿ Applications: Tris solutions are formulated into working concentrations for preparing the stacking and resolving portions of native or SDS-PAGE gels using discontinuous buffer systems according to Laemmli (1970) or Ornstein and Davis (1959). ¿ Use Tris-HCl 1M pH 7.0-7.5 for DEAE Dextran-mediated transfection of non-adherent and adherent cells. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase and Protease Tested ¿ pH Range: 7.5 ¿ 0.1 ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months