Learn More
Teknova 1M Tris HCL, pH 7.5, 500mL, Rnase/Endo Tested, Sterile. 8 pack.
SDP
Catalog No. 502745946
Encompass
1M Tris HCL, pH 7.5, 500mL, Rnase/Endo Tested, Sterile. 8 pack.
Catalog No. 50-274-5946 Supplier Teknova Supplier No. 214668PK
1M Tris HCL, pH 7.5, 500mL, Rnase/Endo Tested, Sterile. 8 pack.