Learn More
Teknova 1M Tris-HCl, pH 7.6 Solution. 1000mL Sterile.
SDP
Catalog No. 502388142
Encompass
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.
Catalog No. 50-238-8142 Supplier Teknova Supplier No. T1076
Add to Cart
Add to Cart
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.