Learn More
Teknova 1M Tris-HCl, pH 8.0 Solution, 20L Bag, Sterile. 4 Pack.
SDP
Catalog No. 502745808
Encompass
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.
Catalog No. 50-274-5808 Supplier Teknova Supplier No. 213454PK
Add to Cart
Add to Cart
Request a Quote
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.