Learn More
Teknova 1M Tris-HCl, pH 8.5. 1000 mL, Sterile.
SDP
Catalog No. 50843270
Encompass
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.
This item is not returnable. View return policy
Catalog No. 50-843-270 Supplier Teknova Supplier No. T1085
Add to Cart
Add to Cart
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.