Learn More
Cayman Chemical 1-methoxy-5-Methylphenazinium (methyl sulfate), Purity: ≥98%, CAS: 65162-13-2, 250 mg
SDP
Catalog No. 502985063
Encompass_Preferred
A photochemically stable mediator of electron transport between NADPH and various electron acceptors such as tetrazolium dyes
Catalog No. 50-298-5063 Supplier Cayman Chemical Supplier No. 21258250 MG
A photochemically stable mediator of electron transport between NADPH and various electron acceptors such as tetrazolium dyes