Learn More
Cayman Chemical 1-Methylhistamine (hydrochloride), Purity: ≥95%, CAS: 6481-48-7, 10 mg
SDP
Catalog No. 502984444
Encompass_Preferred
A major metabolite of histamine used as a biomarker of histaminergic system activity via CSF levels in Alzheimer’s disease patients
Catalog No. 50-298-4444 Supplier Cayman Chemical Supplier No. 1951610 MG
A major metabolite of histamine used as a biomarker of histaminergic system activity via CSF levels in Alzheimer’s disease patients