Learn More
Cayman Chemical 1MethylhIstamInhydrochLR 50mg
SDP
Catalog No. 502983622
Encompass_Preferred
A major metabolite of histamine used as a biomarker of histaminergic system activity via CSF levels in Alzheimer’s disease patients
Catalog No. 50-298-3622 Supplier Cayman Chemical Supplier No. 1951650 MG
A major metabolite of histamine used as a biomarker of histaminergic system activity via CSF levels in Alzheimer’s disease patients