Learn More
Cayman Chemical 1MethylhIstamInhydroCLOr 100mg
SDP
Catalog No. 502984516
Encompass_Preferred
A major metabolite of histamine used as a biomarker of histaminergic system activity via CSF levels in Alzheimer’s disease patients
Catalog No. 50-298-4516 Supplier Cayman Chemical Supplier No. 19516100 MG
A major metabolite of histamine used as a biomarker of histaminergic system activity via CSF levels in Alzheimer’s disease patients