Learn More
Cayman Chemical 1-Methylnicotinamide (chloride), Purity: ≥95%, CAS: 1005-24-9, 100 mg
SDP
Catalog No. 502967531
Encompass_Preferred
An active metabolite of nicotinamide; induces nitric oxide secretion from HUVECs in a concentration-dependent manner; increases plasma levels of 6-keto-prostaglandin F1α in normotensive rats at 30 mg/kg and inhibits collagen-induced thrombus formation in rats at the same dose
Catalog No. 50-296-7531 Supplier Cayman Chemical Supplier No. 16604100 MG
An active metabolite of nicotinamide; induces nitric oxide secretion from HUVECs in a concentration-dependent manner; increases plasma levels of 6-keto-prostaglandin F1α in normotensive rats at 30 mg/kg and inhibits collagen-induced thrombus formation in rats at the same dose