Learn More
Athens Research And Technology Inc 1MG ANTITHROMBIN III HU
Supplier:  Athens Research And Technology Inc 16160120201MG
Encompass_Preferred
Antithrombin III, Human Plasma: 1 MG; Lyophilized from 50 mM Tris-HCl, pH 8.0, with 150 mM NaCl; MW 58,000; EC=0.7; Purity: > =95% by SDS-PAGE; This native protein purified in the Athens Research & Tech lab.
Catalog No. 50-489-624