Learn More
Athens Research And Technology Inc 1MG APOA I HUMAN PLASMA
Supplier:  Athens Research And Technology Inc 16161201011MG
Encompass_Preferred
Apolipoprotein AI, Human Plasma: 1 MG; Frozen in 10 mM NH4HCO3, pH 7.4; MW 28,000; Purity: > =95% by SDS-PAGE; This native protein purified from human plasma in the Athens Research & Technology laboratory. Molecular Biology Reagent, ApoLipoprotein
Catalog No. 50-489-627