Learn More
Abcam Human IgE protein (Azide free), 1MG
Catalog No. NC1824115
Encompass_Preferred
Human IgE protein (Azide free) is a Human Full Length IgE protein with >90% purity, < 25 EU/mg endotoxin level and suitable for western blot, ELISA, IHC, ICC and SDS-PAGE. The predicted molecular weight of ab65866 protein is 185 kDa. - Save time and ensure accurate results - use ab65866 as an IgE isotype control - Optimal bioactivity, stability and reproducibility - No contamination from other immunoglobulin isotypes - Available in different sizes to fit your experimental needs
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin epsilon heavy chain, Immunoglobulin epsilon heavy chain ND
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1824115 Supplier Abcam Supplier No. AB658661MG
Human IgE protein (Azide free) is a Human Full Length IgE protein with >90% purity, < 25 EU/mg endotoxin level and suitable for western blot, ELISA, IHC, ICC and SDS-PAGE. The predicted molecular weight of ab65866 protein is 185 kDa. - Save time and ensure accurate results - use ab65866 as an IgE isotype control - Optimal bioactivity, stability and reproducibility - No contamination from other immunoglobulin isotypes - Available in different sizes to fit your experimental needs
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin epsilon heavy chain, Immunoglobulin epsilon heavy chain ND
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement