Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD2
Supplier:  Biolegend 300240
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD2 [RPA-2.10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Swine (Pig, Porcine); Apps: FC, IHC-F; Size: 1 mg
Catalog No. NC1836480