Learn More
Alkali Scientific 1mL 96-Well Deep Well Plate, V-Bottom, Round Well, Sterile, 5/pk, 100/cs
SDP
Catalog No. 502389474
Encompass_Preferred
Virgin, heat-resistant, medical-grade polypropylene plates. RNase, DNase & pyrogen free. Minimal binding & liquid retention. Black printed alpha-numeric reference grids. V (conical) bottoms. Dimensions: 5.02" L x 3.35" W x 0.96" Hgt. Autoclavable. Temperature range: -86°C to 82.2°C. Sold in full cases only; ten per bag, ten bags per case.
Catalog No. 50-238-9474 Supplier Alkali Scientific Supplier No. MPD32S
Virgin, heat-resistant, medical-grade polypropylene plates. RNase, DNase & pyrogen free. Minimal binding & liquid retention. Black printed alpha-numeric reference grids. V (conical) bottoms. Dimensions: 5.02" L x 3.35" W x 0.96" Hgt. Autoclavable. Temperature range: -86°C to 82.2°C. Sold in full cases only; ten per bag, ten bags per case.