Learn More
Chemglass Life Sciences 1mL Amber Graduated High Recovery Vial with 13-425 Black Phenolic Open Top Cap and Red PTFE/Silicone Septa, 13mm OD x 41mm Height
SDP
Catalog No. 5000980620
Encompass_Preferred
Vials are manufactured from Type 1, Class A Amber Borosilicate glass and come with an open-top cap with PTFE Silicone septa.
Catalog No. 50-009-80620 Supplier Chemglass Life Sciences Supplier No. CG4913A001
Vials are manufactured from Type 1, Class A Amber Borosilicate glass and come with an open-top cap with PTFE Silicone septa.