Learn More
Andwin Scientific 1ML BASE HOLDERS 200/CS
SDP
Catalog No. NC9800876
Encompass_Preferred
The 1 mL base holder is an accessory for the Cyto-Tek Cyto Centrifuge, which is an instrument made to move cells from suspension onto a glass slide by the process of cytocentrifugation. The equipment can hold up to 12 specimen holders. The base holder is sold in a case of 200.
Catalog No. NC9800876 Supplier Andwin Scientific Supplier No. C16609
The 1 mL base holder is an accessory for the Cyto-Tek Cyto Centrifuge, which is an instrument made to move cells from suspension onto a glass slide by the process of cytocentrifugation. The equipment can hold up to 12 specimen holders. The base holder is sold in a case of 200.