Learn More
SAI Infusion Technologies 1.3ml Lithium Heparin Blood Tubes with Flip Cap
SDP
Catalog No. NC2282850
Encompass
1.3ml Lithium Heparin Blood Tube with attached flip cap. Ideal for collection of up to 1.3ml of blood for plasma. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC2282850 Supplier SAI Infusion Technologies Supplier No. PMTPH1.3
1.3ml Lithium Heparin Blood Tube with attached flip cap. Ideal for collection of up to 1.3ml of blood for plasma. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.