Learn More
SAI Infusion Technologies 1.1ml Serum Gel Blood Tubes with Screw Cap
SDP
Catalog No. NC1580159
Encompass
1.1ml Serum Gel Blood Tube with Clotting Activator, Serum Separator Gel and Screw Cap. Ideal for collection of up to 1.1ml of blood for serum. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/box. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC1580159 Supplier SAI Infusion Technologies Supplier No. PMTSSG1.1
1.1ml Serum Gel Blood Tube with Clotting Activator, Serum Separator Gel and Screw Cap. Ideal for collection of up to 1.1ml of blood for serum. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/box. Product is for laboratory animal research only.