Learn More
LifeSensors Control Magnetic Beads (negative control) 1mL
SDP
Catalog No. NC1727817
Encompass_Preferred
For use in “pre-clearing” cell lysates, as well as a negative control for the pulldown of polyubiquitinated proteins in conjunction with magnetic beads-conjugated TUBEs (UM401M and UM402M). Control magnetic beads are generated by blocking an activated matrix (identical to that used for TUBEs conjugation) with ethanolamine. Control magnetic beads (1 mL) are supplied as a slurry in PBS, pH 7.2.
Catalog No. NC1727817 Supplier LifeSensors Supplier No. UM0400M1000
For use in “pre-clearing” cell lysates, as well as a negative control for the pulldown of polyubiquitinated proteins in conjunction with magnetic beads-conjugated TUBEs (UM401M and UM402M). Control magnetic beads are generated by blocking an activated matrix (identical to that used for TUBEs conjugation) with ethanolamine. Control magnetic beads (1 mL) are supplied as a slurry in PBS, pH 7.2.