Learn More
AKADEUM LIFE SCIENCES INC Akadeum DEAD CELL REMOVAL Microbubble Kit
SDP
Catalog No. NC2138291
Encompass
The Dead Cell Removal Microbubble Kit is designed to deplete apoptotic cells from a variety of biological samples. During apoptosis, phosphatidylserine (PS) – a phospholipid normally found on the inner surface of the plasma membrane – is redistributed to the extracellular surface. The relocation of PS to the outer surface of the cell membrane provides an effective target for differentiating dead cells from viable cells within a biological sample. The Dead Cell Removal Kit utilizes BACS™ Microbubbles coated with Annexin V, a PS binding molecule, to capture and float dead cells away. How it Works: The Dead Cell Removal Microbubble Kit are mixed into the sample where they bind to dead and dying cells. Following a centrifugation step, the microbubbles and dead cells are aspirated away while enriched cells are left healthy, untouched, and ready for a wide variety of applications such as single-cell techniques, flow sorting, and cell culturing. Capacity: 1mL
This item is not returnable. View return policy
Catalog No. NC2138291 Supplier AKADEUM LIFE SCIENCES INC Supplier No. 11510211
The Dead Cell Removal Microbubble Kit is designed to deplete apoptotic cells from a variety of biological samples. During apoptosis, phosphatidylserine (PS) – a phospholipid normally found on the inner surface of the plasma membrane – is redistributed to the extracellular surface. The relocation of PS to the outer surface of the cell membrane provides an effective target for differentiating dead cells from viable cells within a biological sample. The Dead Cell Removal Kit utilizes BACS™ Microbubbles coated with Annexin V, a PS binding molecule, to capture and float dead cells away. How it Works: The Dead Cell Removal Microbubble Kit are mixed into the sample where they bind to dead and dying cells. Following a centrifugation step, the microbubbles and dead cells are aspirated away while enriched cells are left healthy, untouched, and ready for a wide variety of applications such as single-cell techniques, flow sorting, and cell culturing. Capacity: 1mL