Learn More
Trajan Scientific 1ML EVOL SYRINGE
SDP
Catalog No. 502530066
Encompass_Preferred
Trajan eVol XCHANGE Syringes, Volume: 1 mL, Length Needle: 50 mm, Needle Point Style: Bevel, Needle Gauge: 23 G, For Use With: Liquid handling procedure, Length: 1.9 in., 50 mm, eVol Syringe
Catalog No. 50-253-0066 Supplier Trajan Scientific Supplier No. 2910035
Trajan eVol XCHANGE Syringes, Volume: 1 mL, Length Needle: 50 mm, Needle Point Style: Bevel, Needle Gauge: 23 G, For Use With: Liquid handling procedure, Length: 1.9 in., 50 mm, eVol Syringe