Learn More
Andwin Scientific 1ML FLUID CHAMBER 50/PK
SDP
Supplier:  Andwin Scientific 6275525
Encompass_Preferred
The 1 mL fluid chamber is an accessory for the Simport CytoSep Funnel Chambers for the Sakura Cyto-Tek Cytocentrifuge. The chamber is sold in a packof 50.
Catalog No. 50-727-569