Learn More
Braintree Scientific 100ml water bottle w/stopper and tube,1ml grad, w/ball
SDP
Catalog No. NC1291655
Encompass
100 ml Glass Water Bottle with stopper and sipper tube. Graduted by one (1) ml. The diameter for the 100ml bottle is 1 1/16" with a 6.5 stopper. Ball point tubeprovides cleaner water by significantly reducing backflow of bacteria into the bottle. Extends bottle use between refills, allows longer cage use between bedding changes and reduces watering costs. Ball Point Tubes have 2 balls and a 5/16 opening. Open drinking tubes (no ball) have aprecision curved tip and stainless steel, thin wall construction.Standard 5/16 diameter with 1/8 end opening. **Sold for research purposes only and not for human use**
Catalog No. NC1291655 Supplier Braintree Scientific Supplier No. WTRBTLSBL
100 ml Glass Water Bottle with stopper and sipper tube. Graduted by one (1) ml. The diameter for the 100ml bottle is 1 1/16" with a 6.5 stopper. Ball point tubeprovides cleaner water by significantly reducing backflow of bacteria into the bottle. Extends bottle use between refills, allows longer cage use between bedding changes and reduces watering costs. Ball Point Tubes have 2 balls and a 5/16 opening. Open drinking tubes (no ball) have aprecision curved tip and stainless steel, thin wall construction.Standard 5/16 diameter with 1/8 end opening. **Sold for research purposes only and not for human use**