Learn More
Analytical Sales & Services 1ML HALF-HGHT 96WELL COLLEC PL
Catalog No. NC2027046
Encompass
1mL Half-Height 96-Well Collection Plate with Shallow V-Bottom Wells, with SiliGuard Low Bind Coating
Catalog No. NC2027046 Supplier Analytical Sales & Services Supplier No. 960101GC
1mL Half-Height 96-Well Collection Plate with Shallow V-Bottom Wells, with SiliGuard Low Bind Coating