Learn More
Trajan Scientific Hamilton 1mL C-line Syringe
Catalog No. NC0931466
Encompass_Preferred
Hamilton Syringe, 1mL, Hamilton C-line Syringe, Fixed Needle, Gas Tight, 22 Gauge, 51mm, Pt. 3
Catalog No. NC0931466 Supplier Trajan Scientific Supplier No. HAM.203081
Hamilton Syringe, 1mL, Hamilton C-line Syringe, Fixed Needle, Gas Tight, 22 Gauge, 51mm, Pt. 3