Learn More
Analytical Sales & Services 1ml Hydrophilic Filter Plate, 0.20µm PP Membrane
Supplier:  Analytical Sales & Services 9615210
Encompass
1Ml Hydrophilic Filter Plate, , Pack/10.
Catalog No. 50-455-972