Learn More
Trajan Scientific L-MARK Brand 1.0mL Syringe, FN, GT, Pt. 3, PTFE Plunger Tip, 22g
Catalog No. 50635272
Encompass_Preferred
L-MARK Brand Syringe, 1mL, Fixed Needle, 22 Gauge, Point Style 3, For Standard LC Adapter, *For use With Headspace Adapter use LMK.2620011 Made in Germany.
Catalog No. 50-635-272 Supplier Trajan Scientific Supplier No. LMK.2620017
L-MARK Brand Syringe, 1mL, Fixed Needle, 22 Gauge, Point Style 3, For Standard LC Adapter, *For use With Headspace Adapter use LMK.2620011 Made in Germany.