Learn More
Trajan Scientific L-MARK Brand 1.0mL Syringe, FN, GT, Pt. 5, PTFE Plunger Tip, 22g
Catalog No. 50635273
Encompass_Preferred
L-MARK Brand 1.0mL, L-MARK Brand Syringe, FN, GT, 22g, NDL Length 57mm, Pt. Style 5, Plunger Tip PTFE, Single Made in Germany.
Catalog No. 50-635-273 Supplier Trajan Scientific Supplier No. LMK.2620019
L-MARK Brand 1.0mL, L-MARK Brand Syringe, FN, GT, 22g, NDL Length 57mm, Pt. Style 5, Plunger Tip PTFE, Single Made in Germany.