Learn More
Analytical Sales & Services 1mL Low Carry Tapered/Round-Bottom Clear Glass Inserts in 96-Well Plate with Blue Pre-Slit Cap Mat
Catalog No. 5000806770
Encompass
1mL Low Carry, Clear Tapered Glass Inserts in 96-Well Sample Prep Plate\n\n• Comes with Blue Opaque Pre-Slit Cap Mat\n\n• Pre-assembled\n\nQty/Pkg: EACH
Catalog No. 50-008-06770 Supplier Analytical Sales & Services Supplier No. 961879
1mL Low Carry, Clear Tapered Glass Inserts in 96-Well Sample Prep Plate\n\n• Comes with Blue Opaque Pre-Slit Cap Mat\n\n• Pre-assembled\n\nQty/Pkg: EACH