Learn More
Chemglass Life Sciences 1mL Micro Volumetric Flask, PTFE Stopper, Tolerance: +-0.01mL
SDP
Catalog No. 50873183
Encompass_Preferred
1mL Micro Volumetric Flask, PTFE Stopper, Tolerance: ¿0.01mL. Similar to CG-1603, but with PTFE stopper.
Catalog No. 50-873-183 Supplier Chemglass Life Sciences Supplier No. CG1603P01
1mL Micro Volumetric Flask, PTFE Stopper, Tolerance: ¿0.01mL. Similar to CG-1603, but with PTFE stopper.