Learn More
Leica Biosystems 1ml NCL-L-CD2-271
Supplier:  Leica Biosystems CD2271LCE
Encompass
Antigen Background The CD2 antigen (LFA-2) is a monomeric 45 to 58 kD glycoprotein. It is an accessory molecule important in mediating the adhesion of activated T cells and thymocytes with antigen-presenting cells and target cells. Disclaimer CD2 (LFA-2) is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2326